3,5-Dimethylphenylboronic acid

Warning

Synonyms: 3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid

480088-5G 480088-25G
220071-331EA 45.14 USD
220071-331 220071-330
3,5-Dimethylphenylboronic acid
3,5-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD00185689
CAS Number: 172975-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR