2,6-Dimethylphenylboronic acid

Synonyms: 2,6-Dimethylbenzeneboronic acid, (2,6-Dimethylphenyl)boronic acid, Dihydroxy(2,6-dimethylphenyl)borane, (2,6-Dimethylphenyl)metaboric acid

480061-25G 480061-5G
220071-327EA 119.77 USD
220071-327 220071-328
2,6-Dimethylphenylboronic acid
2,6-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD01009693
CAS Number: 100379-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR