2,4-Hexadiene mixture of isomers ≥90% (mixture of isomers), Technical Grade

Synonyms: Hexa-2,4-diene

479861-1G 479861-5G 479861-10G
220071-807EA 69.25 USD
220071-807 220071-808 220071-806
2,4-Hexadiene mixture of isomers ≥90% (mixture of isomers), Technical Grade
2,4-Hexadiene
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: -79 °C
Density: 0.7156 g/cm³ (20 °C)
MDL Number: MFCD00009297
CAS Number: 592-46-1
EINECS: 209-757-3
UN: 2458
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR