4-Iodophenyl isocyanate ≥97%

Synonyms: 4-Iodophenylisocyanate

478687-5G 478687-1G
220071-770EA 118.46 USD
220071-770 220071-769
4-Iodophenyl isocyanate ≥97%
4-Iodophenyl isocyanate
Formula: C₇H₄INO
MW: 245.02 g/mol
Boiling Pt: 84 °C (5 -rr)
Melting Pt: 45 - 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013875
CAS Number: 15845-62-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR