2-Bromo-3-hydroxy-4-methoxybenzaldehyde ≥97%

478075-5G 478075-25G
220071-738EA 149.69 USD
220071-738 220071-737
2-Bromo-3-hydroxy-4-methoxybenzaldehyde ≥97%
2-Bromo-3-hydroxy-4-methoxybenzaldehyde
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD02380230
CAS Number: 2973-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR