4-Chloroaniline ≥98%

Danger

477222-1G 477222-10G
220071-662EA 44.58 USD
220071-662 220071-661
4-Chloroaniline ≥98%
4-Chloroaniline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 69 - 71 °C
Density: 1.43 g/cm³ (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00007835
CAS Number: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1,II
Merck Index: 13,02136

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR