2-Hydroxyethyl methacrylate ≥99%

Warning

Synonyms: HEMA, GMA, Methacrylic acid 2-hydroxyethyl ester, 1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate

477028-100ML 477028-25ML
220071-590EA 82.91 USD
220071-590 220071-591
2-Hydroxyethyl methacrylate ≥99%
2-Hydroxyethyl methacrylate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD00002863
CAS Number: 868-77-9
EINECS: 212-782-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR