Formaldehyde diethyl acetal absolute ≥99%

Synonyms: Ethylal, Diethoxymethane

47675-100ML-F 47675-500ML-F
220071-574EA 83.57 USD
220071-574 220069-472
Formaldehyde diethyl acetal absolute ≥99%
Formaldehyde diethyl acetal
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 88 °C (1013 hPa)
Melting Pt: –66 °C
Density: 0.8319 g/cm³ (20 °C)
Flash Pt: –6 °C
Storage Temperature: Ambient
MDL Number: MFCD00009246
CAS Number: 462-95-3
EINECS: 207-330-6
UN: 2373
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR