4-(Trifluoromethyl)phenyl isothiocyanate ≥97%

Synonyms: 1-Isocyanato-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-tolyl isothiocyanate, p-Trifluoromethylphenylisothiocyanate

476099-1G 476099-5G
220071-432EA 63.86 USD
220071-432 220071-433
4-(Trifluoromethyl)phenyl isothiocyanate ≥97%
4-(Trifluoromethyl)phenyl isothiocyanate
Formula: C₈H₄F₃NS
MW: 203.19 g/mol
Boiling Pt: 205…207 °C (1013 hPa)
Melting Pt: 29…31 °C
MDL Number: MFCD00039645
CAS Number: 1645-65-4
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR