1,3-Diiodobenzene ≥98%

Warning

475262-25G 475262-5G
220071-196EA 83.03 USD
220071-196 220071-197
1,3-Diiodobenzene ≥98%
1,3-Diiodobenzene
Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 39 °C
Density: 2.76 g/cm³ (0 °C)
MDL Number: MFCD00041731
CAS Number: 626-00-6
EINECS: 210-921-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR