Barium ≥99.99%

474711-25G 474711-5G
220071-120EA 4261.54 USD
220071-120 220071-121
Barium ≥99.99%
Barium
Formula: Ba
Boiling Pt: 1640 °C (1013 hPa)
Melting Pt: 725 °C
Density: 3.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR