2-Bromophenylboronic acid

Synonyms: (2-Bromophenyl)boronic acid, Dihydroxy(2-bromophenyl)borane, 2-Bromobenzeneboronic acid, (2-Bromophenyl)metaboric acid

473804-5G 473804-25G
220070-311EA 21.05 USD
220070-311 220070-310
2-Bromophenylboronic acid
2-Bromophenylboronic acid
Formula: C₆H₆BBrO₂
MW: 200.83 g/mol
MDL Number: MFCD01114672
CAS Number: 244205-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR