Tungsten hexacarbonyl

472956-5G 472956-25G
220070-982EA 350.77 USD
220070-982 220070-981
Tungsten hexacarbonyl
Tungsten hexacarbonyl
Formula: C₆O₆W
MW: 351.9 g/mol
Boiling Pt: 451.3 °C (1013 hPa)
Melting Pt: 169…171 °C
Density: 2.65 g/cm³ (20 °C)
MDL Number: MFCD00011462
CAS Number: 14040-11-0
EINECS: 237-880-2
UN: 3466
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR