2-Fluoro-5-(trifluoromethyl)phenyl isocyanate ≥97%

Synonyms: 2-Fluoro-5-(trifluoromethyl)phenylisocyanate

472190-2G 472190-500MG
220070-672EA 151.85 USD
220070-672 220070-673
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate ≥97%
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate
Formula: C₈H₃F₄NO
MW: 205.11 g/mol
Boiling Pt: 53 °C (2.5 mmHg)
Density: 1.418 g/cm³ (20 °C)
MDL Number: MFCD00673069
CAS Number: 69922-27-6
UN: 2922
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR