Strontium carbonate ≥99.9%

472018-500G 472018-100G
220070-918EA 315.38 USD
220070-918 220070-917
Strontium carbonate ≥99.9%
Strontium carbonate
Formula: CO₃Sr
MW: 147.63 g/mol
Boiling Pt: 2647 °C (1 mmHg)
Melting Pt: 1768.85 °C
Density: 3.548 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7
Merck Index: 14,08838

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR