4-Iodophenylboronic acid

Synonyms: (4-Iodophenyl)boronic acid, Dihydroxy(4-iodophenyl)borane, 4-Iodobenzeneboronic acid, (4-Iodophenyl)metaboric acid

471933-25G 471933-5G
220070-909EA 245.08 USD
220070-909 220070-910
4-Iodophenylboronic acid
4-Iodophenylboronic acid
Formula: C₆H₆BIO₂
MW: 247.83 g/mol
MDL Number: MFCD01319014
CAS Number: 5122-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR