3,5-Difluorophenylboronic acid

Warning

Synonyms: (3,5-Difluorophenyl)metaboric acid, Dihydroxy(3,5-difluorophenyl)borane, 3,5-Difluorobenzeneboronic acid

471925-25G 471925-5G
220070-906EA 184.85 USD
220070-906 220070-907
3,5-Difluorophenylboronic acid
3,5-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
MDL Number: MFCD01318138
CAS Number: 156545-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR