3,4-Dichlorophenylboronic acid

Warning

471917-25G 471917-5G
220070-904EA 34.62 USD
220070-904 220070-905
3,4-Dichlorophenylboronic acid
3,4-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
MDL Number: MFCD01074646
CAS Number: 151169-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR