α-Bromopropiophenone ≥97%

Synonyms: 2-Bromopropiophenone

471607-5ML 471607-25ML
220070-070EA 41.46 USD
220070-070 220070-069
α-Bromopropiophenone ≥97%
α-Bromopropiophenone
Formula: C₆H₅C(O)CHBrCH₃
MW: 213.07 g/mol
Boiling Pt: 245 °C (1013 hPa)
Density: 1.439 g/cm³ (0 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000145
CAS Number: 2114-00-3
EINECS: 218-307-5
UN: 2927
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR