Dimethyl disulfide ≥99.0%

Danger

Synonyms: Dimethyl disulphide, Methyl disulphide

471569-1L 471569-250ML 471569-25ML
220069-408EA 101.45 USD
220069-408 220069-409 220069-410
Dimethyl disulfide ≥99.0%
Dimethyl disulphide
Formula: C₂H₆S₂
MW: 94.2 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –98 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008561
CAS Number: 624-92-0
EINECS: 210-871-0
UN: 2381
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR