2-(Butylamino)ethanol ≥98%

Synonyms: N-Butylethanolamine

471496-500ML 471496-20L 471496-2L
220070-880EA 75.38 USD
220070-880 220070-878 220070-879
2-(Butylamino)ethanol ≥98%
2-(Butylamino)ethanol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 198…200 °C (1013 hPa)
Density: 0.89 g/cm³ (20 °C)
MDL Number: MFCD00020570
CAS Number: 111-75-1
EINECS: 203-904-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR