2-Ethylaminoethanol ≥98%

Synonyms: N-Ethyl-2-aminoethanol, N-Ethyl-2-ethanolamin, 2-(Ethylamino)ethan-1-ol

471461-500ML 471461-2L 471461-100ML
220070-822EA 61.82 USD
220070-822 220070-821 220070-820
2-Ethylaminoethanol ≥98%
2-Ethylaminoethanol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 167 °C (1013 hPa)
Melting Pt: -9 °C
Density: 0.914 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002841
CAS Number: 110-73-6
EINECS: 203-797-5
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR