2-(Dimethylamino)-ethanol ≥99.5%

Danger

Synonyms: 2-Dimethylaminoethanol, N,N-Dimethylethanolamine, DMAE

471453-18L 471453-500ML 471453-2L 471453-100ML
220070-817EA 592.31 USD
220070-817 220070-819 220070-818 220070-816
2-(Dimethylamino)-ethanol ≥99.5%
2-(Dimethylamino)-ethanol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 132…135 °C (1013 hPa)
Melting Pt: –59 °C
Density: 0.8871 g/cm³ (20 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00002846
CAS Number: 108-01-0
EINECS: 203-542-8
UN: 2051
ADR: 8,II
Merck Index: 13,02863

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR