2-Methylaminoethanol ≥98%

Danger

Synonyms: N-Methyl-2-ethanolamin, N-Methyl-2-aminoethanol, 2-(Methylamino)ethanol

471445-2L 471445-250ML 471445-25ML
220070-025EA 104.03 USD
220070-025 220070-023 220070-024
2-Methylaminoethanol ≥98%
2-Methylaminoethanol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: –3 °C
Density: 0.937 g/cm³ (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00002839
CAS Number: 109-83-1
EINECS: 203-710-0
Merck Index: 12,06096

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR