Methanesulfonic acid ≥99.5%

Danger

471356-500ML 471356-5L 471356-100ML
220070-815EA 97.69 USD
220070-815 220070-006 220070-813
Methanesulfonic acid ≥99.5%
Methanesulphonic acid
Formula: CH₃SO₃H
MW: 96.11 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 189 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 200-898-6
UN: 2586
ADR: 8,III
Merck Index: 13,05981
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebd3f86cbdbb894828247119a403c686b0ecddf&vid=01bb7db9690bd0b27a8ab4f277f8a301da72087f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR