Methanesulfonic acid ≥70% (w/w) in aqueous solution

Danger

471348-100ML 471348-1L 471348-500ML 471348-18L
220070-057EA 30.69 USD
220070-057 220070-059 220070-060 220070-058
Methanesulfonic acid ≥70% (w/w) in aqueous solution
Methanesulphonic acid
Formula: CH₃SO₃H
Storage Temperature: Ambient
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 200-898-6
UN: 2586
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebd3f86cbdbb894828247119a403c686b0ecddf&vid=01bb7db9690bd0b27a8ab4f277f8a301da72087f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR