Isopropylamine ≥99.5%

Danger

Synonyms: 2-Aminopropane

471291-250ML 471291-18L 471291-25ML 471291-2L 471291-1L
220070-040EA 38.12 USD
220070-040 220070-038 220070-041 220070-042 220070-039
Isopropylamine ≥99.5%
Isopropylamine
Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 32.4 °C (1013 hPa)
Melting Pt: –101 °C
Density: 0.69 g/cm³ (20 °C)
Flash Pt: –27 °C
Storage Temperature: Ambient
MDL Number: MFCD00008082
CAS Number: 75-31-0
EINECS: 200-860-9
UN: 1221
ADR: 3,I
Merck Index: 13,05229

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR