Methanesulfonyl chloride ≥99.7%

Danger

Synonyms: Mesyl chloride

471259-500ML 471259-5ML 471259-100ML 471259-1L
220070-808EA 75.46 USD
220070-808 220070-809 220070-806 220070-807
Methanesulfonyl chloride ≥99.7%
Methanesulphonyl chloride
Formula: CH₃ClO₂S
MW: 114.55 g/mol
Boiling Pt: 161 °C (1013 hPa)
Melting Pt: –32 °C
Density: 1.478 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007454
CAS Number: 124-63-0
EINECS: 204-706-1
UN: 3246
ADR: 6.1,I
Merck Index: 13,05982

Order Now

Specification Test Results

Assay 99.7%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR