2,2'-Butyliminodiethanol ≥98.6%

Danger

Synonyms: N-Butyldiethanolamine, N,N-Diethanolbutylamine, N-N-Butyldiethanolamine, N-Butyl-2,2'-iminodiethanol

471240-2L 471240-500ML
220070-139EA 172.31 USD
220070-139 220070-140
2,2'-Butyliminodiethanol ≥98.6%
2,2'-Butyliminodiethanol
Formula: H₃CCH₂CH₂CH₂N(CH₂CH₂OH)₂
MW: 161.24 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.9681 g/cm³ (20 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00002856
CAS Number: 102-79-4
EINECS: 203-055-0

Order Now

Specification Test Results

Assay 98.6%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR