Diethylamine ≥99.5%

Danger

471216-500ML 471216-18L 471216-6X500ML 471216-100ML 471216-1L 471216-5ML
220070-802EA 83.08 USD
220070-802 220070-799 220070-804 220070-798 220070-800 220070-803
Diethylamine ≥99.5%
Diethylamine
Formula: (C₂H₅)₂NH
MW: 73.14 g/mol
Boiling Pt: 55.5 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.7048 g/cm³ (20 °C)
Flash Pt: –39 °C
Storage Temperature: Ambient
MDL Number: MFCD00009032
CAS Number: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: 3,II
Merck Index: 13,03138
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c054fa528239af221505ef08b7c0d2503c9d1d7&vid=e1e59ef071f0c580f9eb9716f4801d314ff824c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR