2,6-Difluorophenylboronic acid ≥98%

Warning

Synonyms: Dihydroxy(2,6-difluorophenyl)borane, 2,6-Difluorobenzeneboronic acid, (2,6-Difluorophenyl)metaboric acid

470791-10G 470791-1G
220070-753EA 162 USD
220070-753 220070-754
2,6-Difluorophenylboronic acid ≥98%
2,6-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
MDL Number: MFCD00792436
CAS Number: 162101-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR