3-((Trifluoromethyl)thio)benzoic acid ≥97%

470090-250MG 470090-1G
220069-768EA 91.11 USD
220069-768 220069-767
3-((Trifluoromethyl)thio)benzoic acid ≥97%
3-((Trifluoromethyl)thio)benzoic acid
Formula: C₈H₅F₃O₂S
MW: 222.19 g/mol
MDL Number: MFCD00236337
CAS Number: 946-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR