2-Chloro-3-(trifluoromethyl)pyridine

469378-5G 469378-25G
220070-567EA 50.18 USD
220070-567 220070-566
2-Chloro-3-(trifluoromethyl)pyridine
2-Chloro-3-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
Boiling Pt: 75…76 °C (30 mmHg)
Melting Pt: 38…39 °C
MDL Number: MFCD00042223
CAS Number: 65753-47-1
EINECS: 424-520-6
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR