3,5-Difluorobenzylamine ≥96%

Danger

469351-5G 469351-1G
220070-565EA 75.17 USD
220070-565 220070-564
3,5-Difluorobenzylamine ≥96%
3,5-Difluorobenzylamine
Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 54 °C (1 mmHg)
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00061244
CAS Number: 90390-27-5
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR