4-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97%

469335-1G 469335-5G
220070-559EA 213.23 USD
220070-559 220070-560
4-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97%
4-Fluoro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
MW: 192.11 g/mol
MDL Number: MFCD00061314
CAS Number: 67515-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR