2,4-Diaminopyrimidine

Warning

Synonyms: Pyrimidine-2,4-diamine, Pyrimidine-2,4-diyltriamine

468231-1G 468231-5G
220070-494EA 109.85 USD
220070-494 220070-495
2,4-Diaminopyrimidine
2,4-Diaminopyrimidine
Formula: C₄H₆N₄
MW: 110.12 g/mol
MDL Number: MFCD00038023
CAS Number: 156-81-0
EINECS: 205-862-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR