(Methyl)triphenylphosphonium chloride

Danger

Synonyms: Methyltriphenylphosphonium chloride, Methyl-TPP-Chloride, MTPP-Chloride

468002-25G 468002-5G
220070-477EA 529.85 USD
220070-477 220070-478
(Methyl)triphenylphosphonium chloride
(Methyl)triphenylphosphonium chloride
Formula: C₁₉H₁₈ClP
MW: 312.78 g/mol
MDL Number: MFCD00797851
CAS Number: 1031-15-8
EINECS: 418-400-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR