5-Bromo-2-thenoic acid

Synonyms: 5-Bromothiophene-2-carboxylic acid, 5-Bromo-2-thiophenecarboxylic acid

467944-5G 467944-1G
220070-472EA 33.92 USD
220070-472 220070-471
5-Bromo-2-thenoic acid
5-Bromo-2-thenoic acid
Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
MDL Number: MFCD00079725
CAS Number: 7311-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR