Lead(II) nitrate ≥99.99%

Danger

Synonyms: Lead dinitrate

467790-50G 467790-250G
220070-455EA 172.31 USD
220070-455 220070-454
Lead(II) nitrate ≥99.99%
Lead(II) nitrate
Formula: Pb(NO₃)₂
MW: 331.21 g/mol
Melting Pt: 450…470 °C
Density: 4.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II
Merck Index: 13,05432

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR