11-Bromo-1-undecene ≥95%

Warning

467642-25ML 467642-5ML
220070-444EA 269.23 USD
220070-444 220070-445
11-Bromo-1-undecene ≥95%
11-Bromo-1-undecene
Formula: C₁₁H₂₁Br
MW: 233.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040825
CAS Number: 7766-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR