5-Bromo-2-fluorobenzonitrile ≥97%

465259-5G 465259-1G
220069-750EA 32.31 USD
220069-750 220069-749
5-Bromo-2-fluorobenzonitrile ≥97%
5-Bromo-2-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 75…80 °C
MDL Number: MFCD00143424
CAS Number: 179897-89-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR