4-Bromo-2-fluorobenzaldehyde ≥96%

Warning

465232-25G 465232-5G
220069-746EA 141.92 USD
220069-746 220069-747
4-Bromo-2-fluorobenzaldehyde ≥96%
4-Bromo-2-fluorobenzaldehyde
Formula: C₇H₄BrFO
MW: 203.01 g/mol
MDL Number: MFCD00143261
CAS Number: 57848-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR