Baicalein

465119-100MG 465119-500MG
220070-200EA 51.69 USD
220070-200 220070-201
Baicalein
Baicalein
Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
MDL Number: MFCD00017459
CAS Number: 491-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR