3,4-Difluorophenylboronic acid

Synonyms: 3,4-Difluorobenzeneboronic acid, (3,4-Difluorophenyl)metaboric acid, (3,4-Difluorophenyl)boronic acid, Dihydroxy(3,4-difluorophenyl)borane

465089-5G 465089-25G
220070-198EA 59.88 USD
220070-198 220070-197
3,4-Difluorophenylboronic acid
3,4-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00807405
CAS Number: 168267-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR