2,4-Difluorophenylboronic acid

Warning

465070-25G 465070-5G
220070-195EA 173.08 USD
220070-195 220070-196
2,4-Difluorophenylboronic acid
2,4-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
MDL Number: MFCD01318998
CAS Number: 144025-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR