5-Hexenoic acid ≥98%

Synonyms: 5-Hexenoicacid

464929-5G 464929-1G
220070-185EA 207.85 USD
220070-185 220070-184
5-Hexenoic acid ≥98%
5-Hexenoic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 107 °C (17 mmHg)
Melting Pt: –37 °C
Density: 0.961 g/cm³ (20 °C)
MDL Number: MFCD00046558
CAS Number: 1577-22-6
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR