Furan-2-boronic acid

Warning

Synonyms: Furan-2-ylboronic acid

464910-10G 464910-1G
220070-182EA 534.46 USD
220070-182 220070-183
Furan-2-boronic acid
Furan-2-boronic acid
Formula: C₄H₅BO₃
MW: 111.89 g/mol
MDL Number: MFCD00799544
CAS Number: 13331-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR