Pyridine-2,6-dicarbonitrile

Synonyms: 2,6-Dicyanopyridine

464899-5G 464899-1G
220070-180EA 356.46 USD
220070-180 220070-179
Pyridine-2,6-dicarbonitrile
Pyridine-2,6-dicarbonitrile
Formula: C₇H₃N₃
MW: 129.12 g/mol
MDL Number: MFCD00129021
CAS Number: 2893-33-6
EINECS: 220-766-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR