(R)-(+)-3-Butyne-2-ol ≥98%

Synonyms: (R)-(+)-But-3-yn-2-ol, (R)-But-3-yn-2-ol, (R)-(+)-3-Hydroxy-1-butyne

464821-5G 464821-1G
220069-465EA 345.69 USD
220069-465 220069-464
(R)-(+)-3-Butyne-2-ol ≥98%
(R)-(+)-3-Butyne-2-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 103 °C (1013 hPa)
Density: 0.891 g/cm³ (20 °C)
MDL Number: MFCD00211237
CAS Number: 42969-65-3
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR