(S)-(-)-3-Butyne-2-ol ≥97%

Synonyms: (S)-3-Butyn-2-ol, (S)-(-)-3-Hydroxy-1-butyne, (S)-(-)-But-3-yn-2-ol

464007-1G 464007-5G
220069-971EA 134.62 USD
220069-971 220069-972
(S)-(-)-3-Butyne-2-ol ≥97%
(S)-(-)-3-Butyne-2-ol
Formula: C₄H₆O
MW: 70.09 g/mol
MDL Number: MFCD00190166
CAS Number: 2914-69-4
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR